Learn More
BD Cell Analysis 3P BD FC Beads APC, Fluorescent Beads for Compensation and Standardization, 5 Tests
Catalog No. NC1383430
Encompass
BD™ FC Beads APC are fluorescent beads designed for precise fluorescence compensation and standardization in flow cytometry applications. They enable accurate spillover value determination and compensation matrix calculation, enhancing performance across BD™ flow cytometers. Ideal for setting up and ensuring consistent results across multiple instruments, these beads offer stability when stored correctly and are intended for research use only.
Catalog No. NC1383430 Supplier BD Cell Analysis 3P Supplier No. 661620
BD™ FC Beads APC are fluorescent beads designed for precise fluorescence compensation and standardization in flow cytometry applications. They enable accurate spillover value determination and compensation matrix calculation, enhancing performance across BD™ flow cytometers. Ideal for setting up and ensuring consistent results across multiple instruments, these beads offer stability when stored correctly and are intended for research use only.