Learn More
BD Cell Analysis 3P BD™ FC Beads FITC for Flow Cytometry Calibration, Fluorescent Beads, Research Use Only, 5 Tests
Catalog No. NC1383413
Encompass
BD™ FC Beads FITC are fluorescent beads designed for flow cytometry calibration and standardization. They ensure accurate voltage settings and fluorescence compensation, enabling reliable multi-color experiments. This product's precision and reliability differentiate it by facilitating consistent results across various BD flow cytometers, essential for research and clinical applications.
Catalog No. NC1383413 Supplier BD Cell Analysis 3P Supplier No. 661615
BD™ FC Beads FITC are fluorescent beads designed for flow cytometry calibration and standardization. They ensure accurate voltage settings and fluorescence compensation, enabling reliable multi-color experiments. This product's precision and reliability differentiate it by facilitating consistent results across various BD flow cytometers, essential for research and clinical applications.