Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fetal Bovine Serum Stain Buffer, BD

Catalog No. BDB554656 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554656 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554656 Supplier BD Biosciences Supplier No. 554656
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the immunofluorescent staining of single-cell suspensions

  • Prepared from lymphoid tissues, bone marrow, peripheral blood,cultured cells
  • Maintain cell viability and maximize fluorescence signal intensities by pH-sensitive fluorochromes
  • Contains metabolic inhibitor, sodium azide
  • For dilution, application of fluorescent reagents and suspension
  • Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Includes Heat-inactivated FBS proteins and less-than-or-equal-to 0.09% sodium azide
Quantity 500mL
Validated Application RUO
Product Type Staining Buffer

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.