Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Fixable Viability Stain 450

Catalog No. BDB562247 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
0.01 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562247 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562247 Supplier BD Supplier No. 562247
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Useful to discriminate viable from non-viable mammalian cells in multicolor flow cytometric application

Permeable plasma cell membranes, such as those present in necrotic cells, allow for the intracellular diffusion of the violet dye and covalent binding to higher overall concentrations of amines than in non-permeable live cells. Therefore, necrotic cells present in a typical in vitro assay label with higher levels of dye increasing their fluorescence intensity 10-20 fold over that of viable cells. The labeled cells can be fixed with formaldehyde for downstream decontamination, freezing and/or permeablization and subsequent intracellular staining while maintaining stable FVS450 fluorescence.

  • Excited by the violet laser (with an excitation maximum of 406nm) and has a fluorescence emission maximum of 450nm
  • Contains a dye that reacts with and covalently binds to cell surface and intracellular amines

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.01 g
For Use With (Application) Research
Storage Requirements -80°C
Product Type Fixable Viability Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.