Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescent Cell Barcoding Wash Buffer, BD

Catalog No. BDB561550 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561550 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561550 Supplier BD Biosciences Supplier No. 561550
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used for washing and staining cells after Fluorescent Cell Barcoding

  • For the dilution and application of fluorescent antibodies to barcoded cells as well as for the suspension, washing, and short-term storage of barcoded cells destined for flow cytometric analysis
  • Formulated as a neutral pH (pH 7.4) -buffered salt solution that is supplemented with fetal bovine serum and bovine serum albumin

Flow Cytometry, Intracellular staining

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorescent cell barcoding Wash Buffer
For Use With (Application) Washing and staining cells after fluorescent cell barcoding, flow cytometry, intracellular staining
Quantity 500 mL
Formulation Neutral pH (pH 7.4) Buffered salt solution, supplemented with Fetal Bovine Serum, Bovine Serum Albumin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.