Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD FoxP3 Staining Kit

Catalog No. BDB560133 Shop All BD Biosciences Products
Change view
Click to view available options
Conjugate:
Alexa Fluor 488
Alexa Fluor 647
PE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
BDB560133 PE
BDB560131 Alexa Fluor 488
BDB560132 Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BDB560133 Supplier BD Supplier No. 560133
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

PE/Alexa Fluor™ 647/ Alexa Fluor 488, FoxP3, CD4, CD25, Buffer set

High performance reagent kit for detection of human FoxP3 positive regulatory T cells

  • Complementing antibody conjugates buffer system fixes and permeabilizes cells in easy, simple steps
  • Components are sufficient for 100 tests
  • Clone: M-A251, RPA-T4, 259D/C7
  • Isotype: mouse IgG1, IgG1, κ, IgG1, κ

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Conjugate PE
Quantity 1 Kit

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.