Learn More
Description
- Isotype: Mouse IgG1, λ
- Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide
Peptide Loading, Immunofluorescent staining, flow cytometric analysis
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | Flow Cytometry, Immunofluorescent Staining, Peptide Loading |
| Formulation | Aqueous Buffered solution containing 0.09% Sodium Azide |
| Name | Mouse H-2Ld:Ig Recombinant Fusion Protein |
| pH Range | 7.2 |
| Purification Method | Affinity Chromatography, SDS-PAGE |
| Quantity | 0.25 mg |
| Storage Requirements | 4°C |
| Recombinant | Recombinant |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.