Learn More
Description
The HLA-A2 protein was expressed together with human β2M in the mouse plasmacytoma cell line, J558L (ATCC TIB-6). The HLA-A2 and β2M polypeptide chains are associated noncovalently as a consequence of their coexpression within J558L cells. The HLA-A2:Ig fusion protein was purified from tissue culture supernatant by affinity chromatography. The purity of the preparation was confirmed by SDS-PAGE.
- Format: Purified
- Isotype: Mouse IgG1, λ
- Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide
- Purified from tissue culture supernatant by affinity chromatography
- Purity was confirmed by SDS-PAGE
Flow cytometry
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | Flow Cytometry |
| Formulation | Aqueous Buffered solution containing 0.09% Sodium Azide |
| Name | Human HLA-A2:Ig Recombinant Fusion Protein |
| pH Range | 7.2 |
| Purification Method | Affinity Chromatography, SDS-PAGE |
| Quantity | 0.05 mg |
| Storage Requirements | 4°C |
| Recombinant | Recombinant |
| Form | Solution |
Research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.