Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD279 (PD-1)

Catalog No. BDB566112 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566112 100 Tests
BDB566175 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566112 Supplier BD Supplier No. 566112
Add to Cart
Add to Cart

CD279 is an immunoregulatory receptor expressed on activated T cells, B cells, and myeloid cells. BD Horizon™ BV480 Mouse Anti-Human CD279 (PD-1) contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms.

With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set

BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510

TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Mouse Anti-Human CD279 (PD-1)
Quantity 100 Tests
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Gene ID (Entrez) 5133
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.