Learn More
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
Specifications
| Product Type | BV480 Mouse Anti-Human CD7 |
| Clonality | Monoclonal |
| Target Species | Human |
| Quantity | 100 Tests |
| For Use With (Application) | Flow Cytometry |
| Cross Reactivity | QC Testing: Human |
| Form | BV480 |
| Sample Volume | 5 μL |
| Content And Storage | Store undiluted at 4°C |
| Gene ID (Entrez) | 924 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.