Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Rat Anti-Human CD197 (CCR7)

Catalog No. BDB566170 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566170 25 Tests
BDB566099 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566170 Supplier BD Supplier No. 566170
Add to Cart
Add to Cart

CCR7 mRNA is expressed mainly in lymphoid tissues including spleen, lymph nodes and tonsil. BD Horizon™ BV480 Rat Anti-Human CD197 (CCR7) is also detected in peripheral T and B lymphocytes, in bone marrow and cord blood CD34-positive cells and mature dendritic cells.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Rat Anti-Human CD197 (CCR7)
Quantity 25 Tests
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.