Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Rat Anti-Mouse CD44

Catalog No. BDB566200 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566200 25 μg
BDB566116 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566200 Supplier BD Supplier No. 566200
Add to Cart
Add to Cart

CD44 is widely expressed on hematopoietic and non-hematopoietic cells. BD Horizon™ BV480 Rat Anti-Mouse CD44 isoforms encoded by variable exons are expressed on epithelial cells, but only at low levels on most leukocytes. Mice with the Ly-24.1 alloantigen (e.g., BALB/c, CBA/J, DBA/1, DBA/2) have relatively large subsets of CD44H+ T lymphocytes.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Rat Anti-Mouse CD44
Quantity 25 μg
For Use With (Application) Flow Cytometry
Concentration 0.2 mg/mL
Cross Reactivity QC Testing: Mouse
Form BV480
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.