Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Rat Anti-Mouse I-A/I-E

Catalog No. BDB566088 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566088 25 μg
BDB566086 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566088 Supplier BD Supplier No. 566088
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M5/114.15.2 monoclonal antibody recognizes a polymorphic determinant shared by I-A[b], I-A[d], I-A[q], I-E[d], I-E[k] MHC class II alloantigens that can be expressed by B cells, dendritic cells, monocytes, macrophages and activated T cells. BD Horizon™ BV480 Rat Anti-Mouse I-A/I-E also reacts with cells from mice of the H-2[p] and H-2[r] haplotypes, and it is non-reactive with cells from NOD (H-2[g7]) mice.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Rat Anti-Mouse I-A/I-E
Quantity 25 μg
For Use With (Application) Flow Cytometry
Concentration 0.2 mg/mL
Cross Reactivity QC Testing: Mouse
Form BV480
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.