Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Horizon™ BV711 Annexin V

Catalog No. BDB563972 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563972 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563972 Supplier BD Supplier No. 563972
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A sensitive probe for identifying apoptotic cells, binding to negatively charged phospholipid surfaces with a higher affinity for phosphatidylserine (PS) than most other phospholipids

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry
Quantity 100 Tests
Color BV711
Content And Storage Store undiluted at 4°C
Form Buffered Solution

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.