Learn More
BD Horizon™ Fixable Viability Stain 510
Catalog No. BDB564406 Shop All Waters Corporation Products
Change view
Click to view available options
Product Type:
Fixable Viability Stain
Catalog No. Product Type
BDB564406 Fixable Viability Stain
1 options
Catalog No. BDB564406 Supplier BD Supplier No. 564406
Add to Cart
Add to Cart
Useful for discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications
  • Reacts with and covalently binds to cell-surface and intracellular amines
  • Permeable plasma cell membranes, such as those present in necrotic cells, allow intracellular diffusion of dye and covalent binding to higher overall concentrations of amines than in non-permeable live cells
  • Necrotic cells present in a typical in vitro assay label with higher levels of dye increases their fluorescence intensity 10-20 fold over viable cells
  • Labeled cells can be fixed with formaldehyde for downstream decontamination, freezing and/or permeabilization and subsequent intracellular staining while maintaining stable viability stain fluorescence
  • Excited by Violet laser with excitation maximum of 408nm and has fluorescence emission maximum of 512nm
Flow cytometry, Intracellular staining
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixable Viability Stain