Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human C3a ELISA Kit

Catalog No. BDB550499 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550499 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550499 Supplier BD Supplier No. 550499
Add to Cart
Add to Cart

For in vitro quantitative determination Human C3a-desArg in human EDTA plasma, serum and other biological samples

The OptEIA ELISA test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay)

  • Utilizes monoclonal antibody specific for human C3a-desArg coated on a 96-well plate

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen C3a
Cross Reactivity Human
Includes Wash concentrate (20X), stop solution, standard/sample diluent, Enzyme concentrate Streptavidin HRP, Human C3a Antibody coated plate, detection Antibody Biotin Anti-Human C3a, Human C3a lyophilized standard, ELISA diluent, TMB one step Substrate reagent
Quantity 1 Plate
Sample Type Human EDTA Plasma, Serum, Biological Sample
Content And Storage 2°C to 8°C
For Use With (Application) In-vitro quantitative determination Human C3a-desArg, ELISA
Product Type Human C3a ELISA kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.