Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human C4a ELISA Kit

Catalog No. BDB550947 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550947 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Toxic, avoid contact with skin, eyes, or clothing
Catalog No. BDB550947 Supplier BD Supplier No. 550947
Add to Cart
Add to Cart
Toxic, avoid contact with skin, eyes, or clothing

Requires

Microplate reader capable of measuring absorbance at 450nm, precision pipettes to deliver 50μL and 100μL volume, adjustable 1mL, 5mL, 10mL, 25mL pipettes for reagent preparation, deionized or distilled water, wash bottle or automated microplate washer, log-log graph paper or automated data reduction, tubes to prepare standard dilutions, laboratory timer, absorbent paper

Quantitative determination of human C4a and C4a-desArg in human EDTA plasma, serum and other biological samples

Utilizes monoclonal antibody specific for human C4a/C4a-desArg coated on 96-well plate. Standards and samples are added to wells, and any C4a-desArg present binds to immobilized antibody. The wells are washed and a mixture of biotinylated polyclonal anti-human C4a antibody and streptavidin-horseradish peroxidase is added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and substrate solution is added, which produces a blue color in direct proportion to the amount of C4a-desArg present in initial sample. Stop Solution changes the color from blue to yellow, wells are read at 450nm

  • Solid phase sandwich ELISA
  • Signals activation of classical or lectin complement pathways in several autoimmune diseases including rheumatoid arthritis, lupus erythematosus, and acute glomerulonephritis as well as in early stages of organ transplant rejections
  • Weak multifunctional proinflammatory mediator by increasing vascular permeability, and to be spasmogenic

TRUSTED_SUSTAINABILITY

Specifications

Antigen C4a
Cross Reactivity Human
Includes Wash concentrate (20X), stop solution, Enzyme concentrate Streptavidin-HRP, ELISA diluent, standard/Sample diluent, TMB one-step Substrate reagent, Human C4a Antibody coated plate, detection Antibody Biotin Anti-Human C4a, Human C4a standard
Quantity 1 Plate
Sample Type Human EDTA Plasma, Serum, Biological Sample
Content And Storage 2°C to 8°C
For Use With (Application) Quantitative determination of Human C4a and C4a-desArg, ELISA
Product Type Human C4a ELISA kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.