Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human C5a ELISA Kit II

Catalog No. BDB557965 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557965 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557965 Supplier BD Supplier No. 557965
Add to Cart
Add to Cart

Includes

Antibody Coated Wells, Detection Antibody, Standards, Enzyme Concentrate (250x), Standard/Sample Diluent, ELISA Diluent, Wash Concentrate (20x), TMB One-Step Substrate Reagent, Stop Solution, Plate Sealers

For the in vitro quantitative determination of Human C5a-desArg in human EDTA plasma, serum and other biological samples

Induces the release of pharmacologically active mediators from a number of cell types

  • Standard Range: 78-5000pg/mL

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen C5a
Cross Reactivity Human
Includes Stop solution, ELISA diluent, TMB one-step Substrate reagent, detection Antibody Biotin Anti-Human C5a, Human C5a standard, wash concentrate (20X), standard/Sample Diluent, Human C5a Antibody coated plate, Enzyme concentrate Streptavidin HRP
Quantity 1 Plate
Sample Type Human EDTA Plasma, Serum, Biological Sample
Content And Storage 2°C to 8°C
For Use With (Application) In-vitro quantitative determination of Human C5a-desArg, ELISA
Product Type Human C5a ELISA kit II

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.