Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human CD253 (TRAIL) ELISA Set

Catalog No. BDB550948 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550948 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Toxic if ingested, inhaled, or in contact with skin
Catalog No. BDB550948 Supplier BD Supplier No. 550948
Add to Cart
Add to Cart
Toxic if ingested, inhaled, or in contact with skin

Includes

  • Capture Antibody: Anti-human CD253 monoclonal antibody
  • Detection Antibody: Biotinylated anti-human CD253 monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant human TRAIL, lyophilized
  • Instruction / Analysis Certificate: (lot-specific)

Requires

96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, microplate reader capable of measuring absorbance at 450nm, precision pipettes, graduated cylinder, one liter, deionized or distilled water, wash bottle or automated washer, log-log graph paper or automated data reduction, tubes to prepare standard dilutions, laboratory timer, plate sealers or parafilm

For human TNF-Related Apoptosis-Inducing Ligand (TRAIL)

  • Alias: TRAIL
  • For natural or recombinant human TRAIL in serum, plasma, and cell culture supernatants

Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY

Specifications

Product Type Human CD253 (TRAIL) ELISA set
Sufficient For 20 x 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Human TRAIL in Serum, Plasma and cell culture supernatants
Kit Contents anti-Human CD253 monoclonal (capture) Antibody, Biotinylated anti-Human CD253 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human TRAIL (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.