Learn More
BD Human Cell Surface Marker Screening Panel
Catalog No. BDB560747 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 Tests
Catalog No. Quantity
BDB560747 5 Tests
1 options
Catalog No. BDB560747 Supplier BD Supplier No. 560747
Includes
Three 96 well plates, each well containing 2.75μg of antibody, enough for five tests (0.5μg/test) along with AlexaFluor™ 647 conjugated goat anti-mouse Ig and goat anti-rat Ig secondary antibodies
For screening cell lines, primary cells or tissue, and is compatible with flow cytometry and bioimaging technology platforms
Bioimaging, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 Tests
Research Use Only