Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Dendritic Cell Enrichment Cocktail, BD

Catalog No. BDB558420 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558420 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Avoid exposure to skin and eyes, ingestion, and contact with heat, acids, and metals
Catalog No. BDB558420 Supplier BD Biosciences Supplier No. 558420
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avoid exposure to skin and eyes, ingestion, and contact with heat, acids, and metals

Includes

  • Human Dendritic Cell Enrichment Cocktail, 5.0 ml:
  • Biotin Mouse Anti-human CD3, clone UCHT1
  • Biotin Mouse Anti-human CD14, clone M5E2
  • Biotin Mouse Anti-human CD19, clone HIB19
  • Biotin Mouse Anti-human CD41a, clone HIP8
  • Biotin Mouse Anti-human CD56, clone B159
  • Biotin Mouse Anti-human CD66b, clone G10F5
  • Biotin Mouse Anti-human CD235a (Glycophorin A), clone GA-R2 (HIR2)
  • Biotin Mouse Anti-human IgE, clone G7-26
  • in Aqueous buffered solution containing 0.09% Sodium Azide
  • BD IMag Streptavidin Particles Plus - DM, 7.5 ml:
  • in Aqueous buffered solution containing BSA™ and 0.09% Sodium Azide

For selecton of negative dendritic cells from peripheral blood

Simultaneously stains erythrocytes, platelets, and most leukocytes except the DC. Negative selection is then performed to enrich for the unlabeled DC

  • Contains biotinylated monoclonal antibodies that recognize antigens expressed on erythrocytes, platelets, and peripheral leukocytes that are not DC
  • BD IMag™ Streptavidin Particles Plus – DM are magnetic nanoparticles that have streptavidin covalently conjugated to their surfaces
  • Avoids the inadvertent activation of enriched DC by using reagents that do not directly bind to DC

Applications:

Flow cytometry, Tissue culture

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Includes Human Dendritic Cell enrichment cocktail, 5mL: Biotin Mouse Anti-Human CD3, clone UCHT1, CD14, clone M5E2, CD19, clone HIB19, CD41a, clone HIP8, CD56, clone B159, CD66b, clone G10F5, CD235a (Glycophorin A), clone GA-R2 (HIR2), IgE, clone G7-26, BD IMag Streptavidin particles Plus - DM, 7.5mL
Quantity 5mL
Validated Application RUO
Product Type Dendritic Cell Enrichment Cocktail

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.