Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human Eotaxin ELISA Set

Catalog No. BDB555175 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555175 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Reagents which contain preservatives may be toxic if ingested, inhaled, or in contact with skin.
Catalog No. BDB555175 Supplier BD Supplier No. 555175
Only null left
Add to Cart
Add to Cart
Reagents which contain preservatives may be toxic if ingested, inhaled, or in contact with skin.

Includes

  • Capture Antibody: Anti-human eotaxin monoclonal antibody
  • Detection Antibody: Biotinylated anti-human eotaxin monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant human eotaxin, lyophilized

Requires

96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, Microplate reader capable of measuring absorbance at 450nm, Precision pipettes, Graduated cylinder, one liter, Deionized or distilled water, Wash bottle or automated washer, Log-log graph paper or automated data reduction, Tubes to prepare standard dilution, Laboratory timer, Plate sealers or parafilm

For natural or recombinant human eotaxin in serum, plasma, and cell culture supernatants

The OptEIA™ Set for human eotaxin contains the components necessary to develop enzyme-linked immunosorbent assays

  • Standard Range: 6.3 - 400pg/mL
  • Calibrated against purified Baculovirus-expressed recombinant human eotaxin
  • Allow flexible assay design to fit individual laboratory needs
  • To design an immunoassay with different sensitivity and dynamic range

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 6.3 to 400pg/mL
Product Type Human Eotaxin ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Human eotaxin in Serum, Plasma and cell culture supernatants
Kit Contents anti-Human eotaxin monoclonal (capture) Antibody, Biotinylated anti-Human eotaxin monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human eotaxin (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.