Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human IFN-γ ELISA Kit II

Catalog No. BDB550612 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550612 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550612 Supplier BD Supplier No. 550612
Add to Cart
Add to Cart

For in vitro quantitative determination of human interferons in serum, plasma, and cell culture samples

The OptEIA™ test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay).

  • Reactivity: Human
  • Utilizes monoclonal antibody specific for IFN-γ coated on a 96-well plate

ELISA

Includes:

  • Antibody Coated Wells: 2 plates of 96 breakable wells (12 strips x 8 wells) coated with anti-human IFN-Γ monoclonal antibody
  • Detection Antibody: 30mL of biotinylated anti-human IFN-Γ monoclonal antibody with ProClin™-150 as preservative
  • Standards: 4 vials lyophilized recombinant human IFN-Γ
  • Enzyme Concentrate (250x): 150μL of 250x concentrated Streptavidin-horseradish peroxidase conjugate with BSA and ProClin-300 as preservative
  • Standard/Sample Diluent: 30mL of animal serum with 0.09% sodium azide as preservative
  • ELISA Diluent: 12mL of a buffered protein base with 0.09% sodium azide as preservative
  • Wash Concentrate (20x): 100mL of 20x concentrated detergent solution with ProClin-150 as preservative
  • TMB One-Step Substrate Reagent: 30 ml of 3,3',5,5'-tetramethylbenzidine (TMB) in buffered solution
  • Stop Solution: 13mL of 1 M phosphoric acid
  • Plate Sealers: 4 sheets with adhesive backing
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN
Cross Reactivity Human
Includes Human IFN-g Antibody coated plate, detection Antibody biotin Anti-Human IFN-g, Human IFN-gamma standard, TMB one-step Substrate reagent, Enzyme concentrate Streptavidin -HRP, wash concentrate (20X), ELISA Diluent, stop solution, standard/Sample Diluent
Quantity 2 Plates
Sample Type Serum, Plasma, Cell Culture
Content And Storage 2°C to 8°C
For Use With (Application) In-vitro quantitative determination of Human interferons, ELISA
Product Type Human IFN-gamma ELISA kit II
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.