Learn More
Description
Includes
Antibody Coated Wells, Detection Antibody, Standards, Enzyme Concentrate (250x), Enzyme Diluent, ELISA Dilution reagent, Wash Concentrate (20x), TMB One-Step Substrate Reagent, Stop Solution, Plate Sealers
- Standard Range: 1.95-125pg/mL
- Potent immunomodulator
- Formulated for improved sensitivity
ELISA
Specifications
Specifications
| Antigen | IL-1beta |
| Conjugate | HRP |
| Cross Reactivity | Human |
| Includes | Stop solution, Human IL-beta standard, TMB one-step Substrate reagent, Enzyme diluent, Human IL-1beta Antibody coated plate, ELISA dilution reagent, detection Antibody Biotin Anti-Human IL-1beta, Enzyme concentrate Streptavidin HRP, wash concentrate (20X) |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| For Use With (Application) | Quantitative determination of Human IL-1beta, ELISA |
| Product Type | Human IL-1beta ELISA kit II |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.