Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human IL-1β Set II

Catalog No. BDB557953 Shop All BD Biosciences Products
Change view
Click to view available options
Concentration:
3.9 to 250pg/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
BDB557953 3.9 to 250pg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557953 Supplier BD Supplier No. 557953
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate

An immunoassay design to fit individual laboratory needs

To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human IL-1β in serum, plasma, and cell culture supernatants

Enzyme-linked immunosorbent assay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 3.9 to 250pg/mL
For Use With (Application) ELISA
Name Human IL-1 beta Set II
Quantity 20 Plates
Storage Requirements -80°C
Cross Reactivity Human

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.