Learn More
Description
Includes
2 plates of 96 breakable wells coated with anti-human IL-10 antibody; Detection antibody: Biotinylated anti-human IL-10; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human IL-10, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers
The OptEIA™ test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay).
- Reactivity: Human
- Utilizes monoclonal antibody specific for IFN-γ coated on a 96-well plate
Applications:
ELISA
Specifications
Specifications
| Antigen | IL-10 |
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Includes | 2 Plates of 96 breakable well coated with Anti-Human IL-10 Antibody, Biotinylated Anti-Human IL-10, HRP Conjugate (250X), 4 vials recombinant Human IL-10 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop solution, adhesive plate sealers |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| For Use With (Application) | Quantitative determination of Human interleukins, ELISA |
| Product Type | Human IL-10 ELISA kit II |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.