Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human IL-12 (p40) ELISA Set

Catalog No. BDB555171 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555171 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Reagents which contain preservatives may be toxic if ingested, inhaled, or in contact with skin
Catalog No. BDB555171 Supplier BD Supplier No. 555171
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagents which contain preservatives may be toxic if ingested, inhaled, or in contact with skin

Includes

  • Capture Antibody: Anti-human IL-12 monoclonal antibody
  • Detection Antibody: Biotinylated anti-human IL-12 monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant human IL-12 (p40), lyophilized

Requires

96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, Microplate reader capable of measuring absorbance at 450nm, Precision pipettes, Graduated cylinder, one liter, Deionized or distilled water, Wash bottle or automated washer, Log-log graph paper or automated data reduction, Tubes to prepare standard dilutions, Laboratory timer, Plate sealers or parafilm

To develop enzyme-linked immunosorbent assays for natural or recombinant human IL-12(p40) in serum, plasma, and cell culture supernatant

BD OptEIA™ Sets allow flexible assay design to fit individual laboratory needs. To design an immunoassay with different sensitivity and dynamic range

  • Standard Range: 31.3 - 2000 pg/ml
  • Calibrated against purified Baculovirus-expressed recombinant human IL-12 (p40)

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 31.3 to 2000pg/mL
Product Type Human IL-12 (p40) ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Human IL-12 (p40) in Serum, Plasma and cell culture supernatant
Kit Contents anti-Human IL-12 monoclonal (capture) Antibody, Biotinylated anti-Human IL-12 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-12 (p40)(standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.