Learn More
Description
Includes
2 plates of 96 breakable wells coated with anti-human IL-12 antibody; Detection antibody: Biotinylated anti-human IL-12; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human IL-12, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers
- Contains biologically active p70 which stimulates the growth of activated CD4+ and CD8+ T cells and NK cells
Specifications
Specifications
| Antigen | IL-12 (p70) |
| Conjugate | Unconjugated |
| Cross Reactivity | Human |
| Includes | 2 Plates of 96 breakable well coated with Anti-Human IL-12 Antibody, Biotinylated Anti-Human IL-12, HRP Conjugate (250X), 4 vials recombinant Human IL-12 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop solution, adhesive plate sealers |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| For Use With (Application) | In-vitro quantitative determination of Human IL-12 (p70), ELISA |
| Product Type | Human IL-12 (p70) ELISA kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.