Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human IL-2 ELISA Kit II

Catalog No. BDB550611 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550611 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550611 Supplier BD Supplier No. 550611
Add to Cart
Add to Cart

Includes

2 plates of 96 breakable wells coated with anti-human IL-2 antibody; Detection antibody: Biotinylated anti-human IL-2; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human IL-2, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers

For quantitative determination of human interleukins in serum, plasma, and cell culture samples

The OptEIA™ test is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay).

  • Utilizes monoclonal antibody specific for IL-2 coated on a 96-well plate
  • Developed for accuracy with serum and plasma specimens

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Cross Reactivity Human
Includes 2 Plates of 96 breakable well coated with Anti-Human IL-2 Antibody, Biotinylated Anti-Human IL-2, HRP Conjugate (250X), 4 vials recombinant Human IL-2 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop
Quantity 2 Plates
Sample Type Serum, Plasma, Cell Culture
Content And Storage 2°C to 8°C
For Use With (Application) Quantitative determination of Human interleukins, ELISA
Product Type Human IL-2 ELISA kit II
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.