Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human IL-2 sRα ELISA Set

Catalog No. BDB559104 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559104 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559104 Supplier BD Supplier No. 559104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Capture Antibody: Anti-human IL-2 sRα, Detection Antibody: Biotinylated anti-human IL-2 sRα, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate, Standard: Recombinant human IL-2 sRα (lyophilized), Instruction/Analysis Certificate

Design an immunoassay with different sensitivity and dynamic range

  • Flexible assay design
  • Calibrated against purified recombinant human IL-2 sRα

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 7.8 to 500pg/mL
Product Type Human IL-2sRa (sCD25) ELISA set
Sufficient For 20 x 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Human IL-2sRa in Serum, Plasma and cell culture supernatants
Kit Contents anti-Human IL-2 sRa monoclonal (capture) Antibody, Biotinylated anti-Human IL-2 sRa monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-2 sRa (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.