Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human IL-4 ELISA Kit II

Catalog No. BDB550614 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550614 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Toxic if ingested, inhaled, or brought in contact with skin
Catalog No. BDB550614 Supplier BD Supplier No. 550614
Add to Cart
Add to Cart
Toxic if ingested, inhaled, or brought in contact with skin

Includes

2 plates of 96 breakable wells coated with anti-human IL-4 antibody; Detection antibody: Biotinylated anti-human IL-4; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human IL-4, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers

Requires

Microplate reader capable of measuring absorbance at 450nm, Precision pipettes to deliver 50μL and 100μL volumes, adjustable 1mL, 5mL, 10mL, 25mL pipettes, deionized or distilled water, wash bottle or automated microplate washer, log-log graph paper or computer and software for ELISA data analysis, Tubes to prepare standard dilutions, laboratory timer, absorbent paper

For quantitative determination of human IL-4 in serum, plasma, and cell culture supernatant

Utilizes monoclonal antibody specific for IL-4 coated on 96-well plate that binds to immobilized antibody. Wells are washed and Streptavidin-horseradish peroxidase conjugate mixed with biotinylated anti-human IL-4 antibody is added, producing antibody-antigen-antibody “sandwich”. Wells are again washed and TMB substrate solution is added, which produces blue color in direct proportion to amount of IL-4 present in initial sample. The stop solution changes the color from blue to yellow, and microwell absorbances are read at 450nm

  • Protein containing 130 amino acid residues
  • Contains standards and samples
  • Produced by activated T cells, mast cells, and basophils
  • Can stimulate the growth, differentiation and maintain viability of certain subsets of B cells and T cells
  • Can have variety of biological effects on many non-lymphoid cells including endothelial cells and fibroblasts
  • Standard Range: 7.8 - 500pg/mL
  • Sensitivity: 4pg/mL

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Cross Reactivity Human
Includes 2 Plates of 96 breakable well coated with Anti-Human IL-4 Antibody, Biotinylated Anti-Human IL-4, HRP Conjugate (250X), 4 vials recombinant Human IL-4 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop
Quantity 2 Plates
Sample Type Serum, Plasma, Cell Culture
Content And Storage 2°C to 8°C
For Use With (Application) Quantitative determination of Human IL-4, ELISA
Product Type Human IL-4 ELISA kit II
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.