Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human IL-8 ELISA Kit II

Catalog No. BDB550999 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550999 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550999 Supplier BD Supplier No. 550999
Add to Cart
Add to Cart

Includes

Antibody coated wells, Detection antibody, Standards, Enzyme Concentrate (250x), Standard/Sample Diluent, ELISA Diluent, Wash Concentrate (20x), TMB One-Step Substrate Reagent, Stop Solution, Plate Sealers

Developed for superior accuracy with serum and plasma specimens

Helps in the quantitative determination of human IL-8 in serum, plasma, and cell culture supernatant.

  • Related to low molecular weight proinflammatory factors known as chemokines

Applications:

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-8
Cross Reactivity Human
Includes Wash concentrate (20X), stop solution, Enzyme concentrate Streptavidin HRP, Human IL-8 Antibody coated plate, detection Antibody Biotin Anti-Human IL-8, recombinant Human IL-8 standard, standard/Sample Diluent, TMB one-step Substrate reagent, ELISA Diluent
Quantity 2 Plates
Sample Type Serum, Plasma
Content And Storage 2°C to 8°C
For Use With (Application) Quantitative determination of Human IL-8, ELISA
Product Type Human IL-8 ELISA kit II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.