Learn More
Description
Includes
BD PharMingen Stain Buffer (FBS); Cytofix/Cytoperm Buffer; Perm/Wash Buffer; PE-conjugated anti-human or mouse IL-2; PE-conjugated anti-human or mouse IFN-g; PE-conjugated anti-human or mouse TNF-a; PE-conjugated isotype control cocktail; Purified blocking antibody cocktail; Leukocyte Activation Cocktail; MiCK-1 or HiCK-1 Cytokine Positive Control Cells; Staining Kit Manual
- Contains the essential reagents for the activation of a human peripheral blood cell population and their subsequent intracellular staining to detect IL-2, IFN-g-, and TNF-a-producing cells
Specifications
Specifications
| Reactivity | Human |
| Detects | IL-2, IFN-g- and TNF-a |
| Quantity | 25 Tests |
| Content And Storage | 4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.