Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human Lineage Cell Depletion Set - DM

Catalog No. BDB560030 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560030 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560030 Supplier BD Supplier No. 560030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Biotin Mouse Anti-human CD3, Biotin Mouse Anti-human CD14, Biotin Mouse Anti-human CD16, Biotin Mouse Anti-human CD19,Biotin Mouse Anti-human CD41a,Biotin Mouse Anti-human CD56, Biotin Mouse Anti-human Glycophorin A, Streptavidin Particles Plus - DM

For depletion of mature hematopoietic cells from PBMC, bone marrow, cord blood samples

For depletion of mature hematopoietic cells from PBMC, bone marrow, cord blood samples

  • Contains biotinylated monoclonal antibodies
  • Enriches rare cell populations such as stem cell, progenitor cells, dendritic cells
  • Contains sufficient reagents to label 1 x 10e9 peripheral blood mononuclear cells
  • Streptavidin Particles Plus - DM are magnetic nanoparticles

Applications:

Cell separation

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Bone Marrow, Cord Blood, PBMC
Type Human Lineage Cell Depletion Set
For Use With (Application) Cell separation
Includes Biotin Mouse Anti-Human CD3, Biotin Mouse Anti-Human CD14, Biotin Mouse Anti-Human CD16, Biotin Mouse Anti-Human CD19, Biotin Mouse Anti-Human CD41a, Biotin Mouse Anti-Human CD56, Biotin Mouse Anti-Human Glycophorin A, Streptavidin particles Plus - DM, Biotinylated monoclonal Antibodies
Quantity 5 mL

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.