Learn More
Description
Includes
2 plates of 96 breakable wells coated with anti-human MCP-1 antibody; Detection antibody: Biotinylated anti-human MCP-1; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human MCP-1, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers
- Solid phase sandwich ELISA
- Utilizes monoclonal antibody specific for MCP-1 coated on a 96-well plate
Applications:
ELISA
Specifications
Specifications
| Antigen | MCP-1 |
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Includes | 2 Plates of 96 breakable well coated with Anti-Human MCP-1 Antibody, Biotinylated Anti-Human MCP-1, HRP Conjugate (250X), 4 vials recombinant Human MCP-1 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop solution, adhesive plate sealers |
| Quantity | 2 Plates |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Content And Storage | 2°C to 8°C |
| Form | Lyophilized |
| For Use With (Application) | Quantitative determination of Human MCP-1, ELISA |
| Product Type | Human MCP-1 ELISA kit |
Safety and Handling
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.