Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human and Mouse Pluripotent Stem Cell Analysis Kit

Catalog No. BDB560477 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560477 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains 4% paraformaldehyde (PFA) and is classified as harmful
Catalog No. BDB560477 Supplier BD Supplier No. 560477
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains 4% paraformaldehyde (PFA) and is classified as harmful

Includes

SSEA-4, Oct3/4, and SSEA-1

To analyze cells for expression of cell surface and intracellular pluripotency markers and differentiation markers

Provides the reagents necessary to perform multicolor flow cytometry on human embryonic stem cells (hESCs), mouse embryonic stem cells (mESCs), and induced pluripotent stem (iPS) cells

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow cytometry
Includes SSEA-4 Alexa Fluor™ 647, Oct3/4 PerCP-Cy™5.5, SSEA-1 PE, isotype control Antibodies, BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD™ compBead Plus compensatio
Quantity 50 Tests

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.