Learn More
BD Human Pluripotent Stem Cell Transcription Factor Analysis Kit
Shop All Waters Corporation ProductsDescription
Includes
Monoclonal antibodies, isotype controls, compensation beads, staining and fixation buffers, verified protocols, and software analysis guidelines
To perform multicolor flow cytometry on human embryonic stem cells (hESCs) and induced pluripotent stem (iPS) cells
- Reactivity: Human
Flow cytometry
Specifications
Specifications
| Formulation | Nanog PE, Oct 3/4 PerCP-Cy™5.5, Sox2 Alexa Fluor™ 647, isotype control Antibodies, BD™ CompBead Plus compensation particles, BD Cytofix™ Fixation Buffer, BD Perm/Wash™ |
| For Use With (Application) | Flow cytometry |
| Includes | Monoclonal Antibodies, isotype controls, compensation beads, Staining and Fixation Buffers, verified protocols and software analysis guidelines |
| Quantity | 50 Tests |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.