Learn More
BD Human Regulatory T Cell Sorting Kit
Catalog No. BDB560753 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB560753 1 Kit
1 options
Catalog No. BDB560753 Supplier BD Supplier No. 560753
Add to Cart
Add to Cart
Includes
Human Regulatory T Cell Sorting Cocktail (CD4 PerCP-Cy5.5, CD25 PE, CD127 Alexa Fluor™ 647), CD45RA FITC, Compensation Controls
Provides reagents that allow investigators to stain viable human peripheral blood mononuclear cells (PBMC)
Flow Cytometry, Fluorescence Activated Cell Sorting
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Research Use Only