Learn More
BD Human T Cell Activation Kit
Catalog No. bdb560750 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB560750 1 Kit
1 options
Catalog No. BDB560750 Supplier BD Supplier No. 560750
Only null left
Add to Cart
Add to Cart
Includes
Alexa Fluor™ 647 Conjugates: p38 MAPK (pT180/Y182), ERK (pT202/204), Stat1 (pY701), Stat3 (pY705), Stat5 (pY694), and Stat6 (pY641) Human T Cell Cocktail, Compensation Reagents, Control Cells and buffers
Reliably determines the level of key phosphorylated signaling proteins involved in T-cell activation
  • Ready to use
  • Supplied with pre-titrated fluorochrome conjugated antibodies, optimized buffers, instrument setup beads, lyophilized human control cells, validated protocols
  • Includes access to Cytobank software for storing, sharing and analysis of data
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For research use only. Not for diagnostic or therapeutic procedures.