Learn More
BD Human Th1/Th17 Phenotyping Kit
Catalog No. BDB560752 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB560752 1 Kit
1 options
Catalog No. BDB560752 Supplier BD Supplier No. 560752
Add to Cart
Add to Cart
Includes
Human Th1/Th17 Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, IFN-γ FITC), BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Simultaneously view Th1 and Th17 cells in mouse model systems
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
  • Can be used to successfully analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For research use only. Not for diagnostic or therapeutic procedures.