Learn More
BD Human Th17/Treg Phenotyping Kit
Catalog No. BDB560762 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB560762 1 Kit
1 options
Catalog No. BDB560762 Supplier BD Supplier No. 560762
Add to Cart
Add to Cart
Includes
Human Th17/Treg Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, FoxP3 Alexa Fluor™ 647), Human FoxP3 Buffer A (10X), Human FoxP3 Buffer B (50X), BD GolgiStop™ Protein Transport Inhibitor
Simultaneously view TH17 and regulatory T cells in human model systems
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
  • Can be used to analyze ex vivo lymphoid cell samples or to monitor CD4+ T cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For research use only. Not for diagnostic or therapeutic procedures.