Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD ICC fixation buffer

Catalog No. BDB550010 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550010 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains formalin, which is harmful and a suspected carcinogen
Catalog No. BDB550010 Supplier BD Supplier No. 550010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains formalin, which is harmful and a suspected carcinogen

For cytokine immunocytochemical staining

Useful for fixation of unstained cells adhered to microscopic slides

  • Neutral pH-buffered saline
  • Used to preserve human and rodent lymphoid cells

Immunocytochemistry (cytospins)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Product Type Fixation Buffer
Quantity 100mL
For Use With (Application) Fixing unstained cells on slides for Cytokine immunocytochemical staining

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.