Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Imag™ Biotin Mouse NK Cell Enrichment Set - DM

Catalog No. bdb557954 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mL Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557954 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Catalog No. BDB557954 Supplier BD Supplier No. 557954
Only null left
Add to Cart
Add to Cart
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

For negative selection of Natural Killer (NK) cells from mouse spleen or lymph node

Mouse NK Cell Enrichment Cocktail contains monoclonal antibodies that recognize antigens expressed on peripheral erythrocytes and leukocytes that are not NK cells. BD™ IMag™ Streptavidin Particles Plus - DM are magnetic nanoparticles that have streptavidin covalently conjugated to their surfaces. With these two components, BD IMag Mouse NK Cell Enrichment Set - DM avoids inadvertent activation of enriched NK cells by using reagents that do not directly bind to those NK cells.

  • Comprises biotin-conjugated monoclonal anti-mouse antibodies: CD4 (L3T4) (clone GK1.5), CD5 (Ly-1) (clone 53-7.3), CD8a (Ly-2) (clone 53-6.7), CD19 (clone 1D3), CD24 (Heat Stable Antigen) (clone M1/69), Ly-6G and Ly-6C (Gr-1) (clone RB6-8C5), TER-119/Erythroid Cells (clone TER-119)
  • Storage buffer: aqueous buffered solution containing BSA and ≤0.09% sodium azide
  • Contains reagents for 109 leukocytes
  • Size: 5mL

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Host Species Mouse
Purification Method Affinity Purified
Quantity 5 mL Each
Regulatory Status RUO
Content And Storage Store undiluted at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.