Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD IMag™ Monocyte Enrichment Set

Catalog No. BDB558454 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mL Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558454 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558454 Supplier BD Supplier No. 558454
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Human Monocyte Enrichment Cocktail
  • Biotin Mouse Anti-Human CD41a
  • Streptavidin Particles Plus - DM

Optimized for used with IMagnet, and contains sufficient reagents to label 10e9 peripheral blood mononuclear cells

Human Monocyte Enrichment Cocktail containing biotinylated monoclonal antibodies recognizes antigens expressed on erythrocytes and peripheral leukocytes.

  • Used for the negative selection of monocytes from peripheral blood

TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Purification Method Affinity Purified
Quantity 5 mL Each
Regulatory Status RUO
Content And Storage Store undiluted at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.