Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Industrial Plate Standard

Catalog No. L99610 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L99610 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L99610 Supplier BD Supplier No. 299610
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tryptic Soy Agar 100mm Plate for general isolation and cultivation of microorganisms.

  • Appearance: Pale white to light yellow and clear to trace hazy
  • Gel Strength: Firm
  • Bioburden: Passed inspection after Seven days
  • Biological Performance: Supports moderate to heavy growth of Escherichia coli ATCC™ 25922, Staphylococcus aureus ATCC™ 25923, and Stenotrophomonas maltophilia13637
  • pH at 25°C: 7.4 (range 7.1 to 7.5)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Country of Origin New Zealand, United States
Form Liquid
Format Plate
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar
Product Type Agar Plate
For Use With (Application) Microbial Cultivation
Incubation Conditions 35 ±2°C
Incubation Time 18 to 48 hr.
Packaging Type Plate
Quantity 100/Pk.
Diameter (Metric) 100 mm
Protein Source Bovine, Porcine
Manufacturing Quality ISO 9001:2008, ISO 13485:2003, FDA Regulated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.