Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Integrins Sampler Kit

Catalog No. BDB611435 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611435 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611435 Supplier BD Supplier No. 611435

Includes

Fibronectin, Integrin a2/VLA-2a, Integrin a3/VLA-3a, Integrin a5, Integrin aL/LFA-1a, Integrin aV / CD51, Integrin b1, Integrin b3, Integrin b4

For routine testing of western blot analysis

  • Format: Purified
  • Storage buffer: Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.