Learn More
BD Interlink™ Vial Access Cannulas
Catalog No. 136897C Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-689-7C 5 mL
13-689-7A 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-689-7C Supplier BD Supplier No. 303403
Add to Cart
Add to Cart
This item is not returnable. View return policy
Interlink System syringes with cannula used for needleless access of a single-dose vial
  • Specifically designed for use with Interlink injection sites
  • Identified by a colored alert ring around the septum
  • BD safety-engineered device
  • Latex-free
Not compatible with conventional injection sites.
IV flush, IV push
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 15 G
For Use With (Application) For needleless access of a single dose vial, IV flush, IV push, for use with Baxter™ Interlink™, Abbott Lifeshield, McGaw Safeline™
Product Type Syringe
Volume (Metric) 5 mL
Connection Type Cannula
Sterility Sterile