Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Leukocyte Activation Cocktail, with BD GolgiPlug™

Catalog No. bdb550583 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 x 100μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550583 2 x 100μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550583 Supplier BD Supplier No. 550583
Only null left
Add to Cart
Add to Cart

Includes

Phorbol ester PMA, calcium ionophore (Ionomycin), protein transport inhibitor BD GolgiPlug™ (brefeldin A)

For elicitating primary cytokine response from T cells

  • Ready-to-use polyclonal cell activation mixture
  • Stimulates cells and produces cytokine
  • Localizes cytokines by protein transport inhibitor component
  • Blocks intracellular protein transport processes
  • Enhances detectability of cytokine-producing cells by immunofluorescent staining and flow cytometric analysis

Intracellular staining

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
pH 7.2
Quantity 2 x 100μL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.