Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Lineage Antibody Cocktail

Catalog No. BDB558074 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558074 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558074 Supplier BD Supplier No. 558074
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

APC Mouse Lineage Antibody Cocktail, APC Mouse Lineage Isotype Control Cocktail

Designed for flow cytometric identification of hematopoietic progenitors in mouse bone marrow

React with cells from the major hematopoietic lineages, such as T lymphocytes, B lymphocytes, monocytes/macrophages, NK cells, erythrocytes, and granulocytes

  • Contains equivalent concentrations of isotype-matched negative-control immunoglobulin
  • Additional fluorochrome-labeled reagents may be combined to further characterize hematopoietic progenitor subpopulations
  • Used to deplete cells bearing hematopoietic lineage markers by flow cytometric or immunomagnetic sorting
  • 20μl of antibody or isotype control cocktail is sufficient to stain each sample of 106 leukocytes
  • Excited by laser lines from 595 to 647nm, and its emission is collected in a detector for fluorescence wavelengths between 640 and 680nm

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Conjugate APC
Host Species Mouse
Purification Method Affinity Purified
Quantity 1 Each
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.