Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Liquid Amies Elution Swab (Eswab) Collection/Transport System
GSA_VA

Catalog No. 22349701 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Tip Type:
Flexible Minitip Flocked
Minitip Flocked
Regular Flocked
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Type
22-349-701 Minitip Flocked
22-349-700 Regular Flocked
B220532 Flexible Minitip Flocked
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-349-701 Supplier BD Supplier No. 220246
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Polypropylene screw-cap tube filled with 1mL of Liquid Amies transport medium and specimen collection swab

Intended for the collection and transport of clinical specimens containing aerobes, anaerobes and fastidious bacteria from the collection site to the testing laboratory

One of the routine procedures in the diagnosis of bacteriological infections involves the collection and safe transportation of swab samples. This can be accomplished using the BD Liquid Amies Elution Swab (ESwab) Collection and Transport System. BD ESwab incorporates a modified Liquid Amies transporting medium, which can sustain the viability of a plurality of organisms that include clinically important aerobes, anaerobes and fastidious bacteria such as Neisseria gonorrhoeae during transit to the testing laboratory.

  • Processed using standard clinical laboratory operating procedures for bacterial culture
  • Swab tip flocked with soft nylon fiber
  • Supplied sterile in a collection kit format with labeled conical-bottom tube

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Elution Swab Collection Kit
Media Type Liquid Amies
Sterility Sterile
Swab Configuration Single Applicator
Closure Color Green
Fill Volume 1 mL
For Use With (Application) For the collection and transport of clinical specimens containing aerobes, anaerobes and fastidious Bacteria
Includes Includes Polypropylene screw-cap tube with 1mL of liquid Amies medium with a Green cap, one minitip swab with flocked Nylon Fiber tip
Quantity 50 Pk.
Storage Requirements 5°C to 25°C
Tip Type Minitip Flocked
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.